missing translation for 'onlineSavingsMsg'
Learn More

D-Valine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. V00945G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00945G 5 g
V00941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V00945G Supplier TCI America Supplier No. V00945G
Only null left

Chemical Identifiers

CAS 7146-15-8
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.63
MDL Number MFCD00237309
InChI Key KUGLDBMQKZTXPW-NUBCRITNSA-N
Synonym d-valine methyl ester hydrochloride, methyl d-valinate hydrochloride, h-d-val-ome.hcl, r-methyl 2-amino-3-methylbutanoate hydrochloride, d-valinemethyl ester hydrochloride, methyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, methyl ester, hydrochloride, h-d-val-ome, methyl d-valinate hcl, h-d-val-ome hcl
PubChem CID 11984198
IUPAC Name methyl (2R)-2-amino-3-methylbutanoate hydrochloride
SMILES Cl.COC(=O)[C@H](N)C(C)C

Specifications

Melting Point 165°C
Color White
Quantity 5 g
Formula Weight 167.63
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material D-Valine Methyl Ester Hydrochloride