missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Valine Methyl Ester Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 165°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 167.63 |
| Percent Purity | ≥98.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | D-Valine Methyl Ester Hydrochloride |
Safety and Handling
| TSCA | No |