missing translation for 'onlineSavingsMsg'
Learn More

D-Valine ethyl ester hydrochloride, 97%

Catalog No. AAH6661903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6661903 1 g
AAH6661906 5 g
AAH6661914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6661903 Supplier Thermo Scientific Chemicals Supplier No. H6661903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 73913-64-1 | C7H16ClNO2 | 181.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73913-64-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00191043
InChI Key PQGVTLQEKCJXKF-FYZOBXCZSA-N
Synonym ethyl d-valinate hydrochloride, r-ethyl 2-amino-3-methylbutanoate hydrochloride, d-valine ethyl ester hydrochloride, h-d-val-oet.hcl, ethyl d-valinate hcl, ethyl 2r-2-amino-3-methylbutanoate hydrochloride, h-d-val-oet?cl, h-d-val-oet. hcl, ethyld-valinatehydrochloride, d-valine ethylester hydrochloride
PubChem CID 42609611
IUPAC Name ethyl (2R)-2-amino-3-methylbutanoate;hydrochloride
SMILES CCOC(=O)C(C(C)C)N.Cl

Specifications

Melting Point 102°C to 104°C
Beilstein 3912412
Quantity 1 g
Formula Weight 181.66
Percent Purity 97%
Physical Form Solid
Chemical Name or Material D-Valine ethyl ester hydrochloride
EINECSNumber 277-635-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only