accessibility menu, dialog, popup

UserName

CAS RN 73913-64-1

H3COOCH3CH3NH2HCl

CAS RN 73913-64-1

IUPAC Name: ethyl 2-amino-3-methylbutanoate hydrochloride
Synonyms: ethyl 2-amino-3-methylbutanoate hydrochloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: PQGVTLQEKCJXKF-UHFFFAOYNA-N
SMILES: Cl.CCOC(=O)C(N)C(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)