missing translation for 'onlineSavingsMsg'
Learn More

D-Valine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. V00955G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00955G 5 g
V00951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V00955G Supplier TCI America Supplier No. V00955G

Chemical Identifiers

CAS 17662-84-9
Molecular Formula C19H25NO5S
Molecular Weight (g/mol) 379.471
MDL Number MFCD00136545
InChI Key QWUQVUDPBXFOKF-RFVHGSKJSA-N
Synonym H-D-Val-OBzl.Tos-OH
PubChem CID 44629854
IUPAC Name benzyl (2R)-2-amino-3-methylbutanoate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.CC(C)C(C(=O)OCC1=CC=CC=C1)N

Specifications

Melting Point 162°C
Color White
Quantity 5 g
Formula Weight 379.47
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Valine Benzyl Ester p-Toluenesulfonate