accessibility menu, dialog, popup

UserName

CAS RN 17662-84-9

IUPAC Name: 4-methylbenzene-1-sulfonic acid; benzyl 2-amino-3-methylbutanoate
Synonyms: benzyl 2-amino-3-methylbutanoate; para-toluene sulfonate
Molecular Weight (g/mol): 379.47
Molecular Formula: C19H25NO5S
InChi Key: QWUQVUDPBXFOKF-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.CC(C)C(N)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results