missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Tyrosine 98.0+%, TCI America™
$79.74 - $233.52
Chemical Identifiers
| CAS | 556-02-5 |
|---|---|
| Molecular Formula | C9H11NO3 |
| Molecular Weight (g/mol) | 181.19 |
| MDL Number | MFCD00063073 |
| InChI Key | OUYCCCASQSFEME-SVGMAFHSNA-N |
| Synonym | d-tyrosine, h-d-tyr-oh, d-tyr, r-2-amino-3-4-hydroxyphenyl propanoic acid, d-tyrosin, 2r-2-amino-3-4-hydroxyphenyl propanoic acid, 3-4-hydroxyphenyl-d-alanine, d-p-tyrosine, tyrosine, d, d-+-tyrosine |
| PubChem CID | 71098 |
| ChEBI | CHEBI:28479 |
| IUPAC Name | (2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid |
| SMILES | N[C@H](CC1=CC=C(O)C=C1)C(O)=O |
Chemical Identifiers
| 556-02-5 | |
| 181.19 | |
| OUYCCCASQSFEME-SVGMAFHSNA-N | |
| 71098 | |
| (2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid |
| C9H11NO3 | |
| MFCD00063073 | |
| d-tyrosine, h-d-tyr-oh, d-tyr, r-2-amino-3-4-hydroxyphenyl propanoic acid, d-tyrosin, 2r-2-amino-3-4-hydroxyphenyl propanoic acid, 3-4-hydroxyphenyl-d-alanine, d-p-tyrosine, tyrosine, d, d-+-tyrosine | |
| CHEBI:28479 | |
| N[C@H](CC1=CC=C(O)C=C1)C(O)=O |
Specifications
| 314°C | |
| C9H11NO3 | |
| d-tyrosine, h-d-tyr-oh, d-tyr, r-2-amino-3-4-hydroxyphenyl propanoic acid, d-tyrosin, 2r-2-amino-3-4-hydroxyphenyl propanoic acid, 3-4-hydroxyphenyl-d-alanine, d-p-tyrosine, tyrosine, d, d-+-tyrosine | |
| N[C@H](CC1=CC=C(O)C=C1)C(O)=O | |
| 5 g | |
| 71098 | |
| 181.19 | |
| Crystalline Powder |
| 556-02-5 | |
| MFCD00063073 | |
| OUYCCCASQSFEME-SVGMAFHSNA-N | |
| (2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid | |
| 181.19 | |
| CHEBI:28479 | |
| ≥98.0% (T) | |
| D-Tyrosine |
Safety and Handling
EINECSNumber : (9)-1596
TSCA : Yes