Learn More
D-Tyrosine methyl ester hydrochloride, 98%
CAS: 3728-20-9 | C10H14ClNO3 | 231.68 g/mol
$94.08 - $358.36
Chemical Identifiers
| CAS | 3728-20-9 |
|---|---|
| Molecular Formula | C10H14ClNO3 |
| Molecular Weight (g/mol) | 231.68 |
| MDL Number | MFCD00066124 |
| InChI Key | VXYFARNRGZWHTJ-MVRPGTNWNA-N |
| Synonym | h-d-tyr-ome.hcl, d-tyrosine methyl ester hydrochloride, h-d-tyr-ome hcl, methyl d-tyrosinate hydrochloride, r-methyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, d-tyrosine, methyl ester, hydrochloride, methyl 2r-2-amino-3-4-hydroxyphenyl propanoate hydrochloride, methyl d-tyrosinate hcl, tyrosine methylester hydrochloride, h-d-tyr-omehcl |
| PubChem CID | 134565 |
| IUPAC Name | methyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride |
| SMILES | Cl.COC(=O)[C@H](N)CC1=CC=C(O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6363006
|
Thermo Scientific Chemicals
H6363006 |
5 g |
N/A
|
|
|||||
|
AAH6363014
|
Thermo Scientific Chemicals
H6363014 |
25 g |
N/A
|
|
|||||
Description
D-Tyrosine methyl ester hydrochloride is an intermediate in the synthesis of oxybutynin chloride (O868525), an inhibitor of proliferation and supresses gene expression in bladder smooth muscle cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Tyrosine methyl ester hydrochloride is an intermediate in the synthesis of oxybutynin chloride (O868525), an inhibitor of proliferation and supresses gene expression in bladder smooth muscle cells.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
| 3728-20-9 | |
| 231.68 | |
| VXYFARNRGZWHTJ-MVRPGTNWNA-N | |
| 134565 | |
| Cl.COC(=O)[C@H](N)CC1=CC=C(O)C=C1 |
| C10H14ClNO3 | |
| MFCD00066124 | |
| h-d-tyr-ome.hcl, d-tyrosine methyl ester hydrochloride, h-d-tyr-ome hcl, methyl d-tyrosinate hydrochloride, r-methyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, d-tyrosine, methyl ester, hydrochloride, methyl 2r-2-amino-3-4-hydroxyphenyl propanoate hydrochloride, methyl d-tyrosinate hcl, tyrosine methylester hydrochloride, h-d-tyr-omehcl | |
| methyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride |
Specifications
| 3728-20-9 | |
| C10H14ClNO3 | |
| h-d-tyr-ome.hcl, d-tyrosine methyl ester hydrochloride, h-d-tyr-ome hcl, methyl d-tyrosinate hydrochloride, r-methyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, d-tyrosine, methyl ester, hydrochloride, methyl 2r-2-amino-3-4-hydroxyphenyl propanoate hydrochloride, methyl d-tyrosinate hcl, tyrosine methylester hydrochloride, h-d-tyr-omehcl | |
| Cl.COC(=O)[C@H](N)CC1=CC=C(O)C=C1 | |
| 5 g | |
| 134565 | |
| 98% | |
| D-Tyrosine methyl ester hydrochloride |
| White | |
| MFCD00066124 | |
| VXYFARNRGZWHTJ-MVRPGTNWNA-N | |
| methyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride | |
| 231.68 | |
| 231.68 | |
| Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 223-084-2
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only