accessibility menu, dialog, popup

UserName

CAS RN 3728-20-9

IUPAC Name: methyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate hydrochloride
Synonyms: methyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate hydrochloride
Molecular Weight (g/mol): 231.68
Molecular Formula: C10H14ClNO3
InChi Key: VXYFARNRGZWHTJ-MVRPGTNWNA-N
SMILES: Cl.COC(=O)[C@H](N)CC1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)