missing translation for 'onlineSavingsMsg'
Learn More

D-Tyrosine methyl ester hydrochloride, 98%

Catalog No. AAH6363006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6363006 5 g
AAH6363014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6363006 Supplier Thermo Scientific Chemicals Supplier No. H6363006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3728-20-9 | C10H14ClNO3 | 231.68 g/mol

D-Tyrosine methyl ester hydrochloride is an intermediate in the synthesis of oxybutynin chloride (O868525), an inhibitor of proliferation and supresses gene expression in bladder smooth muscle cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Tyrosine methyl ester hydrochloride is an intermediate in the synthesis of oxybutynin chloride (O868525), an inhibitor of proliferation and supresses gene expression in bladder smooth muscle cells.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 3728-20-9
Molecular Formula C10H14ClNO3
Molecular Weight (g/mol) 231.68
MDL Number MFCD00066124
InChI Key VXYFARNRGZWHTJ-MVRPGTNWNA-N
Synonym h-d-tyr-ome.hcl, d-tyrosine methyl ester hydrochloride, h-d-tyr-ome hcl, methyl d-tyrosinate hydrochloride, r-methyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, d-tyrosine, methyl ester, hydrochloride, methyl 2r-2-amino-3-4-hydroxyphenyl propanoate hydrochloride, methyl d-tyrosinate hcl, tyrosine methylester hydrochloride, h-d-tyr-omehcl
PubChem CID 134565
IUPAC Name methyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](N)CC1=CC=C(O)C=C1

Specifications

Color White
Quantity 5 g
Formula Weight 231.68
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Tyrosine methyl ester hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-084-2
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only