missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ D-Tyrosine, 98%

$452.97

Chemical Identifiers

CAS 556-02-5
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00063073
InChI Key OUYCCCASQSFEME-MRVPVSSYSA-N
Synonym d-tyrosine, h-d-tyr-oh, d-tyr, r-2-amino-3-4-hydroxyphenyl propanoic acid, d-tyrosin, 2r-2-amino-3-4-hydroxyphenyl propanoic acid, 3-4-hydroxyphenyl-d-alanine, d-p-tyrosine, tyrosine, d, d-+-tyrosine
PubChem CID 71098
ChEBI CHEBI:28479
IUPAC Name (2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES N[C@H](CC1=CC=C(O)C=C1)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC226910100
View Documents Promotion Details
Thermo Scientific™
226910100
10g Glass bottle
Each for $452.97
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

556-02-5
181.19
OUYCCCASQSFEME-MRVPVSSYSA-N
71098
(2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid
C9H11NO3
MFCD00063073
d-tyrosine, h-d-tyr-oh, d-tyr, r-2-amino-3-4-hydroxyphenyl propanoic acid, d-tyrosin, 2r-2-amino-3-4-hydroxyphenyl propanoic acid, 3-4-hydroxyphenyl-d-alanine, d-p-tyrosine, tyrosine, d, d-+-tyrosine
CHEBI:28479
N[C@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

556-02-5
0.5% max. (105°C, 3 hrs)
C9H11NO3
MFCD00063073
14, 605
14, 9839
OUYCCCASQSFEME-MRVPVSSYSA-N
(2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid
181.19
CHEBI:28479
≥97.5%
Authentic
98%
4-(HO)C6H4CH2CH(NH2)CO2H
0.1% max.
+10° to +12° (20°C, 589nm) (c=5, 1N HCl) (after 2 hrs.)
d-tyrosine, h-d-tyr-oh, d-tyr, r-2-amino-3-4-hydroxyphenyl propanoic acid, d-tyrosin, 2r-2-amino-3-4-hydroxyphenyl propanoic acid, 3-4-hydroxyphenyl-d-alanine, d-p-tyrosine, tyrosine, d, d-+-tyrosine
N[C@H](CC1=CC=C(O)C=C1)C(O)=O
+11.30° (20°C c=5,1N HCl)
71098
181.19
D-Tyrosine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

RUO – Research Use Only