missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Turanose 98.0+%, TCI America™

Catalog No. p-7136336
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T05421G 1 g
T05425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T05421G Supplier TCI America Supplier No. T05421G

Chemical Identifiers

CAS 547-25-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006606
InChI Key RULSWEULPANCDV-UHFFFAOYNA-N
Synonym d +-turanose
PubChem CID 131632488
IUPAC Name 1,4,5,6-tetrahydroxy-3-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexan-2-one
SMILES OCC(O)C(O)C(OC1OC(CO)C(O)C(O)C1O)C(=O)CO

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 342.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Turanose