accessibility menu, dialog, popup

UserName

HOHOOHOOOHOHHOHOOOH

CAS RN 547-25-1

IUPAC Name: 1,4,5,6-tetrahydroxy-3-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexan-2-one
Synonyms: turanose
Molecular Weight (g/mol): 342.3
Molecular Formula: C12H22O11
InChi Key: RULSWEULPANCDV-UHFFFAOYNA-N
SMILES: OCC(O)C(O)C(OC1OC(CO)C(O)C(O)C1O)C(=O)CO
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)