missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Turanose 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Blanc-jaune |
| Quantity | 1 g |
| Formula Weight | 342.30 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | D-(+)-Turanose |
Sécurité et manipulation
| TSCA | Yes |