missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Turanose, 98%

Catalog No. AAB2122406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2122406 5 g
AAB2122403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2122406 Supplier Thermo Scientific Chemicals Supplier No. B2122406

CAS: 547-25-1 | C12H22O11 | 342.30 g/mol

D-Turanose is used as a carbon source by bacteria, fungi, and other organisms. It is involved in intracellular sugar signaling in plant cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Turanose is used as a carbon source by bacteria, fungi, and other organisms. It is involved in intracellular sugar signaling in plant cells.

Solubility
Soluble in water and methanol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in a well sealed container. Store at room temperature.

Chemical Identifiers

CAS 547-25-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006606
InChI Key RULSWEULPANCDV-UHFFFAOYNA-N
Synonym d +-turanose
PubChem CID 131632488
IUPAC Name (3S,4S,5R)-2-(hydroxymethyl)-3-[(2R,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-2,4,5-triol
SMILES OCC(O)C(O)C(OC1OC(CO)C(O)C(O)C1O)C(=O)CO

Specifications

Melting Point ∼170°C (decomposition)
Quantity 5 g
Beilstein 93771
Merck Index 14,9821
Solubility Information Soluble in water and methanol.
Formula Weight 342.3
Percent Purity 98%
Chemical Name or Material D-Turanose
EINECSNumber 208-918-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only