missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Trehalose, Dihydrate, cGMP Manufactured, 97-102.%, Spectrum™ Chemical
T1724, 6138-23-4, C12H22O11·2H2O
$9,859.09 - $178,847.03
Chemical Identifiers
| CAS | 6138-23-4 |
|---|---|
| Molecular Formula | C12H26O13 |
| Molecular Weight (g/mol) | 378.33 |
| MDL Number | MFCD00071594 |
| InChI Key | DPVHGFAJLZWDOC-DJCWUSJTNA-N |
| IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate |
| SMILES | O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18602005
|
N/A | 1 kg | Poly Bottle |
Each for $9,859.09
|
|
||||
|
18602006
|
N/A | 12 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18602007
|
N/A | 25 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical D-(+)-Trehalose, Dihydrate, cGMP Manufactured - Spectrum cGMP grade products that have been manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 6138-23-4 | |
| 378.33 | |
| DPVHGFAJLZWDOC-DJCWUSJTNA-N | |
| O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
| C12H26O13 | |
| MFCD00071594 | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate |
Specifications
| 6138-23-4 | |
| C12H26O13 | |
| +197 to +201° | |
| O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O | |
| 378.33 | |
| Reagent |
| 100% | |
| MFCD00071594 | |
| DPVHGFAJLZWDOC-DJCWUSJTNA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate | |
| 97 to 102.% |