missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Trehalose, Dihydrate, cGMP Manufactured, 97-102.%, Spectrum™ Chemical

Catalog No. 18602005
Change view
Click to view available options
Quantity:
1 kg
12 kg
25 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602005 1 kg Poly Bottle
18602006 12 kg Poly Bottle
18602007 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18602005 Supplier Spectrum Chemical Mfg Cor Supplier No. T17241KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

T1724, 6138-23-4, C12H22O11·2H2O

Spectrum™ Chemical D-(+)-Trehalose, Dihydrate, cGMP Manufactured - Spectrum cGMP grade products that have been manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H26O13
Molecular Weight (g/mol) 378.33
MDL Number MFCD00071594
InChI Key DPVHGFAJLZWDOC-DJCWUSJTNA-N
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
SMILES O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 1 kg
Specific Rotation +197 to +201°
Percent Purity 97 to 102.%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material D-(+)-Trehalose, Dihydrate