Learn More
Thermo Scientific Chemicals D-(+)-Trehalose dihydrate, 99%
CAS: 6138-23-4 | C12H22O11 | 342.30 g/mol
$63.00 - $569.38
Chemical Identifiers
| CAS | 6138-23-4 |
|---|---|
| Molecular Formula | C12H22O11 |
| Molecular Weight (g/mol) | 342.30 |
| MDL Number | MFCD00006628 |
| InChI Key | HDTRYLNUVZCQOY-LBYRQFQKNA-N |
| Synonym | trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose |
| PubChem CID | 7427 |
| ChEBI | CHEBI:16551 |
| IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol |
| SMILES | OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAA1943406
|
Thermo Scientific Chemicals
A1943406 |
5 g |
Each for $63.00
|
|
|||||
|
AAA1943414
|
Thermo Scientific Chemicals
A1943414 |
25 g |
Each for $183.12
|
|
|||||
|
AAA1943422
|
Thermo Scientific Chemicals
A1943422 |
100 g |
Each for $569.38
|
|
|||||
Description
D-(+)-Trehalose dihydrate is used as a food additive in many processed foods and pharmaceutical excipients. It is widely used as a direct replacement of conventional sweeteners and protects the cells from various environmental stresses. It finds application in the treatment of Huntington's disease and Parkinson's disease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water and hot alcohol.
Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 6138-23-4 | |
| 342.30 | |
| HDTRYLNUVZCQOY-LBYRQFQKNA-N | |
| 7427 | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol |
| C12H22O11 | |
| MFCD00006628 | |
| trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose | |
| CHEBI:16551 | |
| OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
Specifications
| 6138-23-4 | |
| 0.99 | |
| MFCD00006628 | |
| Hygroscopic | |
| Soluble in water and hot alcohol. | |
| OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O | |
| 342.30 | |
| CHEBI:16551 | |
| D-(+)-Trehalose dihydrate |
| 97°C to 99°C | |
| C12H22O11 | |
| 5322018 | |
| trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose | |
| HDTRYLNUVZCQOY-LBYRQFQKNA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol | |
| 7427 | |
| 378.33 (342.29 Anhydrous) |
Safety and Handling
H500
EINECSNumber : 202-739-6
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only