missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Trehalose dihydrate, 99%

Catalog No. AAA1943406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1943406 5 g
AAA1943414 25 g
AAA1943422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1943406 Supplier Thermo Scientific Chemicals Supplier No. A1943406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6138-23-4 | C12H22O11 | 342.30 g/mol

D-(+)-Trehalose dihydrate is used as a food additive in many processed foods and pharmaceutical excipients. It is widely used as a direct replacement of conventional sweeteners and protects the cells from various environmental stresses. It finds application in the treatment of Huntington's disease and Parkinson's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and hot alcohol.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006628
InChI Key HDTRYLNUVZCQOY-LBYRQFQKNA-N
Synonym trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose
PubChem CID 7427
ChEBI CHEBI:16551
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 97°C to 99°C
Assay Percent Range 0.99
Quantity 5 g
Beilstein 5322018
Sensitivity Hygroscopic
Solubility Information Soluble in water and hot alcohol.
Formula Weight 378.33 (342.29 Anhydrous)
Chemical Name or Material D-(+)-Trehalose dihydrate
Hazard Category H500
EINECSNumber 202-739-6
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only