missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Trehalose dihydrate, 99%

CAS: 6138-23-4 | C12H26O13 | 378.33 g/mol

Supplier:  Thermo Scientific Chemicals 182550025

 View more versions of this product

Catalog No. AC182550025

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D(+)-Trehalose dihydrate
97.0°C to 99.0°C
Plastic Bottle
MFCD00071594
159742
+ 179.00
(10% in water) Clear colorless
O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
378.33
378.32
6138-23-4
98.5% min. (ELSD) (HPLC)
C12H26O13
2.5 kg
+ 179.00 (20.00°C c=2,H2O)
trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol
DPVHGFAJLZWDOC-DJCWUSJTNA-N
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol;dihydrate
181978
99%

Chemical Identifiers

6138-23-4
378.33
DPVHGFAJLZWDOC-DJCWUSJTNA-N
181978
O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
C12H26O13
MFCD00071594
trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol;dihydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

May form combustible dust concentrations in air

Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only