Learn More
Thermo Scientific Chemicals D-(+)-Trehalose, anhydrous
CAS: 99-20-7 | C12H22O11 | 342.30 g/mol
$66.67 - $372.56
Chemical Identifiers
| CAS | 99-20-7 |
|---|---|
| Molecular Formula | C12H22O11 |
| Molecular Weight (g/mol) | 342.30 |
| MDL Number | MFCD00006628 |
| InChI Key | HDTRYLNUVZCQOY-LBYRQFQKNA-N |
| Synonym | trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose |
| PubChem CID | 7427 |
| ChEBI | CHEBI:16551 |
| IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol |
| SMILES | OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6600606
|
Thermo Scientific Chemicals
J6600606 |
5 g |
Each for $66.67
|
|
|||||
|
AAJ6600609
|
Thermo Scientific Chemicals
J6600609 |
10 g |
Each for $93.49
|
|
|||||
|
AAJ6600618
|
Thermo Scientific Chemicals
J6600618 |
50 g |
Each for $372.56
|
|
|||||
Description
Trehalose, Dihydrate, is an inducer of autophagy, it protect cells against numerous environmental stresses. It is seldom used as a direct replacement for conventional sweeteners. Used as a cryoprotectant in a variety of cell freezing media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTrehalose, Dihydrate, is an inducer of autophagy, it protect cells against numerous environmental stresses. It is seldom used as a direct replacement for conventional sweeteners. Used as a cryoprotectant in a variety of cell freezing media.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
| 99-20-7 | |
| 342.30 | |
| HDTRYLNUVZCQOY-LBYRQFQKNA-N | |
| 7427 | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol |
| C12H22O11 | |
| MFCD00006628 | |
| trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose | |
| CHEBI:16551 | |
| OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
Specifications
| 99-20-7 | |
| C12H22O11 | |
| 5 g | |
| trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose | |
| HDTRYLNUVZCQOY-LBYRQFQKNA-N | |
| OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O | |
| 342.30 | |
| CHEBI:16551 | |
| Powder |
| White | |
| MFCD00006628 | |
| Hygroscopic | |
| Soluble in water. | |
| +197° (c=7 in Water) | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol | |
| 7427 | |
| 342.3 | |
| D-(+)-Trehalose, anhydrous |
Safety and Handling
EINECSNumber : 202-739-6
RTECSNumber : LZ5776770
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only