missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Trehalose, 99%, anhydrous

Catalog No. AC309870250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC309870250 25 g Plastic bottle
AC309871000 100 g Plastic bottle
AC309875000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC309870250 Supplier Thermo Scientific Chemicals Supplier No. 309870250

CAS: 99-20-7 | C12H22O11 | 342.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-20-7
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006628
InChI Key HDTRYLNUVZCQOY-LBYRQFQKNA-N
Synonym trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose
PubChem CID 7427
ChEBI CHEBI:16551
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 203.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) on dry basis (HPLC)
Packaging Plastic bottle
Quantity 25 g
Merck Index 15,9742
Specific Rotation Condition +197.50 (20.00°C c=2,H2O)
Specific Rotation + 197.50
Solubility Information (10% in water) Clear colorless to light yellow
Formula Weight 342.3
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material D-Trehalose, Anhydrous
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-739-6

RUO – Research Use Only