missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-α-Tocotrienol, ≥97% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Vitamins, Nutraceuticals and Natural Products
Specifications
Specifications
| CAS | 58864-81-6 |
| Physical Form | Neat |
| Linear Formula | C29H44O2 |
| MDL Number | MFCD11045308 |
| Synonym | (R)-alpha-Tocotrienol; [R-(E,E)]-3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2 H-1-benzopyran-6-ol |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 424.66 |
| Percent Purity | ≥97% (HPLC) |
| Grade | Analytical Standard |