missing translation for 'onlineSavingsMsg'
Learn More

D-delta-Tocopherol, 93%

Catalog No. AC421051000
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421051000 100 mg Glass bottle
AC421055000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC421051000 Supplier Thermo Scientific Chemicals Supplier No. 421051000

CAS: 119-13-1 | C27H46O2 | 402.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119-13-1
Molecular Formula C27H46O2
Molecular Weight (g/mol) 402.66
MDL Number MFCD20486794
InChI Key GZIFEOYASATJEH-VHFRWLAGSA-N
Synonym delta-tocopherol, unii-r0zb2556p8 component, unii-zl0rfa4e5n component, s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-2h-1-benzopyran-6-ol, 2s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-1-benzopyran-6-ol, 2h-1-benzopyran-6-ol,3,4-dihydro-2,8-dimethyl-2-4r,8r-4,8,12-trimethyltridecyl-, 2r
PubChem CID 12444418
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=CC(O)=CC(C)=C2O1

Specifications

Color Brown to Yellow
Flash Point >93°C
Infrared Spectrum Authentic
Assay Percent Range 91% min. (GC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 15,9656
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol, acetone, chloroform, ether,, oils, fats
IUPAC Name (2S)-2,8-dimethyl-2-[(4S,8S)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
Formula Weight 402.65
Percent Purity 93%
Physical Form Viscous Liquid
Chemical Name or Material D-δ-Tocopherol
Show More Show Less
EINECSNumber 204-299-

RUO – Research Use Only