missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Threonine 98.0+%, TCI America™
Supplier: TCI America T0228100G
Specifications
| D-(+)-Threonine | |
| White | |
| MFCD00064269 | |
| d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dth | |
| C[C@H](O)[C@@H](N)C(O)=O | |
| 119.12 | |
| CHEBI:16398 | |
| ≥98.0% (T) |
| 632-20-2 | |
| C4H9NO3 | |
| 100 g | |
| AYFVYJQAPQTCCC-STHAYSLISA-N | |
| (2R,3S)-2-amino-3-hydroxybutanoic acid | |
| 69435 | |
| 119.12 | |
| Crystalline Powder |
Chemical Identifiers
| 632-20-2 | |
| 119.12 | |
| AYFVYJQAPQTCCC-STHAYSLISA-N | |
| 69435 | |
| (2R,3S)-2-amino-3-hydroxybutanoic acid |
| C4H9NO3 | |
| MFCD00064269 | |
| d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dth | |
| CHEBI:16398 | |
| C[C@H](O)[C@@H](N)C(O)=O |
Safety and Handling
EINECSNumber : (9)-1584
RTECSNumber : XO8580000
TSCA : No