accessibility menu, dialog, popup

UserName

H3C(S)OH(R)NH2OHO

CAS RN 632-20-2

IUPAC Name: (2R,3S)-2-amino-3-hydroxybutanoic acid
Synonyms: D-threonine
Molecular Weight (g/mol): 119.12
Molecular Formula: C4H9NO3
InChi Key: AYFVYJQAPQTCCC-STHAYSLISA-N
SMILES: C[C@H](O)[C@@H](N)C(O)=O
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)