missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Threonine Benzyl Ester Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 130°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 245.70 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | D-Threonine Benzyl Ester Hydrochloride |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |