missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Threonine, 98%

Catalog No. AC346970050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC346970050 5 g Glass Bottle
AC346970250 25 g Glass Bottle
AC346971000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC346970050 Supplier Thermo Scientific Chemicals Supplier No. 346970050
Only null left

CAS: 632-20-2 | C4H9NO3 | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 632-20-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064269
InChI Key AYFVYJQAPQTCCC-STHAYSLISA-N
Synonym d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dth
PubChem CID 69435
ChEBI CHEBI:16398
IUPAC Name (2R,3S)-2-amino-3-hydroxybutanoic acid
SMILES C[C@H](O)[C@@H](N)C(O)=O

Specifications

Ammonium 200ppm max.
Arsenic (As) 1ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 0.3% max.
Assay Percent Range 98%
Other Amino Acids (TLC) Passes Test
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH(NH)COH
Specific Rotation +27° to +29° (20°C, 589nm) (c=6, water)
Sulfate 300ppm max.
Transmittance 98% min. (c=5, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Specific Rotation Condition +28° (20°C c=6,water)
Quantity 5 g
Formula Weight 119.12
Percent Purity ≥97.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D-Threonine
Show More Show Less

RUO – Research Use Only