missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-tert-Leucine 98.0+%, TCI America™
$55.92 - $165.90
Chemical Identifiers
| CAS | 26782-71-8 |
|---|---|
| Molecular Formula | C6H13NO2 |
| Molecular Weight (g/mol) | 131.18 |
| MDL Number | MFCD00004265 |
| InChI Key | NPDBDJFLKKQMCM-UHFFFAOYNA-N |
| Synonym | d-tert-leucine, h-d-tle-oh, d-tert-butylglycine, r-2-amino-3,3-dimethylbutanoic acid, d-t-leucine, r-tert-leucine, 2r-2-amino-3,3-dimethylbutanoic acid, d-t-butylglycine, h-tbu-d-gly-oh, d-valine, 3-methyl |
| PubChem CID | 6950340 |
| IUPAC Name | 2-amino-3,3-dimethylbutanoic acid |
| SMILES | CC(C)(C)C(N)C(O)=O |
Chemical Identifiers
| 26782-71-8 | |
| 131.18 | |
| NPDBDJFLKKQMCM-UHFFFAOYNA-N | |
| 6950340 | |
| CC(C)(C)C(N)C(O)=O |
| C6H13NO2 | |
| MFCD00004265 | |
| d-tert-leucine, h-d-tle-oh, d-tert-butylglycine, r-2-amino-3,3-dimethylbutanoic acid, d-t-leucine, r-tert-leucine, 2r-2-amino-3,3-dimethylbutanoic acid, d-t-butylglycine, h-tbu-d-gly-oh, d-valine, 3-methyl | |
| 2-amino-3,3-dimethylbutanoic acid |
Specifications
| 26782-71-8 | |
| C6H13NO2 | |
| 1 g | |
| NPDBDJFLKKQMCM-UHFFFAOYNA-N | |
| 2-amino-3,3-dimethylbutanoic acid | |
| 6950340 | |
| ≥98.0% (T) | |
| D-tert-Leucine |
| White | |
| MFCD00004265 | |
| d-tert-leucine, h-d-tle-oh, d-tert-butylglycine, r-2-amino-3,3-dimethylbutanoic acid, d-t-leucine, r-tert-leucine, 2r-2-amino-3,3-dimethylbutanoic acid, d-t-butylglycine, h-tbu-d-gly-oh, d-valine, 3-methyl | |
| CC(C)(C)C(N)C(O)=O | |
| 131.18 | |
| 131.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No