missing translation for 'onlineSavingsMsg'
Learn More

D-tert-Leucine 98.0+%, TCI America™

Catalog No. L01575G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01575G 5 g
L01571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01575G Supplier TCI America Supplier No. L01575G

Chemical Identifiers

CAS 26782-71-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00004265
InChI Key NPDBDJFLKKQMCM-UHFFFAOYNA-N
Synonym d-tert-leucine, h-d-tle-oh, d-tert-butylglycine, r-2-amino-3,3-dimethylbutanoic acid, d-t-leucine, r-tert-leucine, 2r-2-amino-3,3-dimethylbutanoic acid, d-t-butylglycine, h-tbu-d-gly-oh, d-valine, 3-methyl
PubChem CID 6950340
IUPAC Name 2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)C(N)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 131.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-tert-Leucine