missing translation for 'onlineSavingsMsg'
Learn More

D-tert-Leucine, 99%

Catalog No. p-7046369
Change view
Click to view available options
Quantity:
500 mg
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14098ME 500 mg
AAL1409804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14098ME Supplier Thermo Scientific Chemicals Supplier No. L14098ME
Only null left

CAS: 26782-71-8 | C6H13NO2 | 131.18 g/mol

D-tert-Leucine is used for synthesis of amino acids e.g. in the synthesis of aldehydes by double alkylation of unsaturated aldimines of tert-Leu esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-tert-Leucine is used for synthesis of amino acids e.g. in the synthesis of aldehydes by double alkylation of unsaturated aldimines of tert-Leu esters.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 26782-71-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00004265
InChI Key NPDBDJFLKKQMCM-UHFFFAOYNA-N
Synonym d-tert-leucine, h-d-tle-oh, d-tert-butylglycine, r-2-amino-3,3-dimethylbutanoic acid, d-t-leucine, r-tert-leucine, 2r-2-amino-3,3-dimethylbutanoic acid, d-t-butylglycine, h-tbu-d-gly-oh, d-valine, 3-methyl
PubChem CID 6950340
IUPAC Name (2R)-2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)C(N)C(O)=O

Specifications

Melting Point >300°C
Beilstein 1721825
Solubility Information Soluble in water.
Quantity 500 mg
Formula Weight 131.18
Optical Rotation −7° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material D-tert-Leucine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only