missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Serine, 98%
CAS: 312-84-5 | C3H7NO3 | 105.093 g/mol
Chemical Identifiers
| CAS | 312-84-5 |
|---|---|
| Molecular Formula | C3H7NO3 |
| Molecular Weight (g/mol) | 105.093 |
| MDL Number | MFCD00004269 |
| InChI Key | MTCFGRXMJLQNBG-UWTATZPHSA-N |
| Synonym | d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin |
| PubChem CID | 71077 |
| ChEBI | CHEBI:16523 |
| IUPAC Name | (2R)-2-amino-3-hydroxypropanoic acid |
| SMILES | C(C(C(=O)O)N)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC227070250
|
N/A | 25 g | Glass Bottle |
N/A
|
|
||||
|
AC227071000
|
N/A | 100 g | Glass Bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C3H7NO3 | |
| MFCD00004269 | |
| d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin | |
| CHEBI:16523 | |
| C(C(C(=O)O)N)O |
Specifications
| 220.0°C | |
| 98% | |
| HOCH2CH(NH2)CO2H | |
| − 14.75 | |
| Solubility in water: soluble in water: 346g/L (20°C). Other solubilities: insoluble in alcohol, ether and benzene., soluble in mineral acids., insoluble in organic solvents. | |
| C(C(C(=O)O)N)O | |
| −14.75° (20°C c=10 2 N HCl) | |
| 71077 | |
| 105.09 | |
| D-Serine |
| 312-84-5 | |
| C3H7NO3 | |
| MFCD00004269 | |
| d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin | |
| MTCFGRXMJLQNBG-UWTATZPHSA-N | |
| (2R)-2-amino-3-hydroxypropanoic acid | |
| 105.093 | |
| CHEBI:16523 | |
| 98% |
RUO – Research Use Only