missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Serine, 98%
CAS: 312-84-5 | C3H7NO3 | 105.093 g/mol
Supplier: Thermo Scientific Chemicals 227070250
| Quantity | 25 g |
|---|---|
| Packaging | Glass Bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C3H7NO3 | |
| MFCD00004269 | |
| d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin | |
| CHEBI:16523 | |
| C(C(C(=O)O)N)O |
Specifications
| 220.0°C | |
| 312-84-5 | |
| Glass Bottle | |
| HOCH2CH(NH2)CO2H | |
| − 14.75 | |
| Solubility in water: soluble in water: 346g/L (20°C). Other solubilities: insoluble in alcohol, ether and benzene., soluble in mineral acids., insoluble in organic solvents. | |
| C(C(C(=O)O)N)O | |
| −14.75° (20°C c=10 2 N HCl) | |
| 105.093 | |
| CHEBI:16523 | |
| 98% |
| D-Serine | |
| 98% | |
| C3H7NO3 | |
| MFCD00004269 | |
| d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin | |
| MTCFGRXMJLQNBG-UWTATZPHSA-N | |
| (2R)-2-amino-3-hydroxypropanoic acid | |
| 25 g | |
| 71077 | |
| 105.09 |