missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Serine, 98%
CAS: 312-84-5 | C3H7NO3 | 105.093 g/mol
Supplier: Thermo Scientific Chemicals 227071000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 220.0°C | |
| 312-84-5 | |
| C3H7NO3 | |
| HOCH2CH(NH2)CO2H | |
| − 14.75 | |
| Solubility in water: soluble in water: 346g/L (20°C). Other solubilities: insoluble in alcohol, ether and benzene., soluble in mineral acids., insoluble in organic solvents. | |
| C(C(C(=O)O)N)O | |
| −14.75° (20°C c=10 2 N HCl) | |
| 105.093 | |
| CHEBI:16523 | |
| 98% |
| D-Serine | |
| 98% | |
| Glass Bottle | |
| MFCD00004269 | |
| d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin | |
| MTCFGRXMJLQNBG-UWTATZPHSA-N | |
| (2R)-2-amino-3-hydroxypropanoic acid | |
| 100 g | |
| 71077 | |
| 105.09 |
Chemical Identifiers
| 312-84-5 | |
| 105.093 | |
| MTCFGRXMJLQNBG-UWTATZPHSA-N | |
| 71077 | |
| (2R)-2-amino-3-hydroxypropanoic acid |
| C3H7NO3 | |
| MFCD00004269 | |
| d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin | |
| CHEBI:16523 | |
| C(C(C(=O)O)N)O |
RUO – Research Use Only