missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Ribono-1,4-lactone 97.0+%, TCI America™
$108.21 - $315.16
Chemical Identifiers
| CAS | 5336-08-3 |
|---|---|
| Molecular Formula | C5H8O5 |
| Molecular Weight (g/mol) | 148.11 |
| MDL Number | MFCD00063241 |
| InChI Key | CUOKHACJLGPRHD-UHFFFAOYNA-N |
| Synonym | ribonolactone, d-ribono-1,4-lactone, d-ribonolactone, ribono-gamma-lactone, d +-ribonic acid gamma-lactone, d-+-ribono-1,4-lactone, d-ribopentono-1,4-lactone, d-ribono-gamma-lactone, d-+-ribonic acid gamma-lactone, d-ribonic acid-1,4-lactone |
| PubChem CID | 111064 |
| ChEBI | CHEBI:74168 |
| IUPAC Name | (3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one |
| SMILES | C(C1C(C(C(=O)O1)O)O)O |
Chemical Identifiers
| 5336-08-3 | |
| 148.11 | |
| CUOKHACJLGPRHD-UHFFFAOYNA-N | |
| 111064 | |
| (3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one |
| C5H8O5 | |
| MFCD00063241 | |
| ribonolactone, d-ribono-1,4-lactone, d-ribonolactone, ribono-gamma-lactone, d +-ribonic acid gamma-lactone, d-+-ribono-1,4-lactone, d-ribopentono-1,4-lactone, d-ribono-gamma-lactone, d-+-ribonic acid gamma-lactone, d-ribonic acid-1,4-lactone | |
| CHEBI:74168 | |
| C(C1C(C(C(=O)O1)O)O)O |
Specifications
| 5336-08-3 | |
| C5H8O5 | |
| 1 g | |
| CUOKHACJLGPRHD-UHFFFAOYNA-N | |
| (3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one | |
| 111064 | |
| 148.11 | |
| Crystalline Powder |
| 86°C | |
| MFCD00063241 | |
| ribonolactone, d-ribono-1,4-lactone, d-ribonolactone, ribono-gamma-lactone, d +-ribonic acid gamma-lactone, d-+-ribono-1,4-lactone, d-ribopentono-1,4-lactone, d-ribono-gamma-lactone, d-+-ribonic acid gamma-lactone, d-ribonic acid-1,4-lactone | |
| C(C1C(C(C(=O)O1)O)O)O | |
| 148.11 | |
| CHEBI:74168 | |
| ≥97.0% (GC) | |
| D-(+)-Ribono-1,4-lactone |
Safety and Handling
TSCA : No