missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Ribono-1,4-lactone 97.0+%, TCI America™

Catalog No. R00631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00631G 1 g
R00635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R00631G Supplier TCI America Supplier No. R00631G

Chemical Identifiers

CAS 5336-08-3
Molecular Formula C5H8O5
Molecular Weight (g/mol) 148.11
MDL Number MFCD00063241
InChI Key CUOKHACJLGPRHD-UHFFFAOYNA-N
Synonym ribonolactone, d-ribono-1,4-lactone, d-ribonolactone, ribono-gamma-lactone, d +-ribonic acid gamma-lactone, d-+-ribono-1,4-lactone, d-ribopentono-1,4-lactone, d-ribono-gamma-lactone, d-+-ribonic acid gamma-lactone, d-ribonic acid-1,4-lactone
PubChem CID 111064
ChEBI CHEBI:74168
IUPAC Name (3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one
SMILES C(C1C(C(C(=O)O1)O)O)O

Specifications

Melting Point 86°C
Quantity 1 g
Formula Weight 148.11
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Ribono-1,4-lactone