missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Ribonic acid gamma-lactone, 96%

Catalog No. AC227830100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC227830100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC227830100 Supplier Thermo Scientific Chemicals Supplier No. 227830100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5336-08-3 | C5H8O5 | 148.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5336-08-3
Molecular Formula C5H8O5
Molecular Weight (g/mol) 148.12
MDL Number MFCD00063241
InChI Key CUOKHACJLGPRHD-UHFFFAOYNA-N
Synonym ribonolactone, d-ribono-1,4-lactone, d-ribonolactone, ribono-gamma-lactone, d +-ribonic acid gamma-lactone, d-+-ribono-1,4-lactone, d-ribopentono-1,4-lactone, d-ribono-gamma-lactone, d-+-ribonic acid gamma-lactone, d-ribonic acid-1,4-lactone
PubChem CID 111064
ChEBI CHEBI:74168
IUPAC Name (3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one
SMILES C(C1C(C(C(=O)O1)O)O)O

Specifications

Melting Point 83.0°C to 85.0°C
Ignition Residue 0.2% max
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 10 g
Specific Rotation Condition + 18.00 (20.00°C c=5,H2O)
Specific Rotation + 18.00
Solubility Information Solubility in water: soluble
Formula Weight 148.12
Percent Purity 96%
Chemical Name or Material D(+)-Ribonic acid γ-lactone, 96%
Show More Show Less
EINECSNumber 226-256-5

RUO – Research Use Only