missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Ribonic acid-1,4-lactone
CAS: 5336-08-3 | C5H8O5 | 148.11 g/mol
Supplier: Thermo Scientific Chemicals J6626606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| D-Ribonic acid-1,4-lactone | |
| 85°C to 87°C | |
| C5H8O5 | |
| 5 g | |
| ribonolactone, d-ribono-1,4-lactone, d-ribonolactone, ribono-gamma-lactone, d +-ribonic acid gamma-lactone, d-+-ribono-1,4-lactone, d-ribopentono-1,4-lactone, d-ribono-gamma-lactone, d-+-ribonic acid gamma-lactone, d-ribonic acid-1,4-lactone | |
| CUOKHACJLGPRHD-UHFFFAOYNA-N | |
| (3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one | |
| 111064 | |
| 148.11 |
| 5336-08-3 | |
| White to Yellow | |
| MFCD00063241 | |
| 82057 | |
| Soluble in water at 100mg/ml | |
| C(C1C(C(C(=O)O1)O)O)O | |
| 148.11 | |
| CHEBI:74168 | |
| Powder |
Chemical Identifiers
| 5336-08-3 | |
| 148.11 | |
| CUOKHACJLGPRHD-UHFFFAOYNA-N | |
| 111064 | |
| (3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one |
| C5H8O5 | |
| MFCD00063241 | |
| ribonolactone, d-ribono-1,4-lactone, d-ribonolactone, ribono-gamma-lactone, d +-ribonic acid gamma-lactone, d-+-ribono-1,4-lactone, d-ribopentono-1,4-lactone, d-ribono-gamma-lactone, d-+-ribonic acid gamma-lactone, d-ribonic acid-1,4-lactone | |
| CHEBI:74168 | |
| C(C1C(C(C(=O)O1)O)O)O |
Safety and Handling
EINECSNumber : 226-256-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only