missing translation for 'onlineSavingsMsg'
Learn More

D-Ribonic acid-1,4-lactone

Catalog No. AAJ6626603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6626603 1 g
AAJ6626606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6626603 Supplier Thermo Scientific Chemicals Supplier No. J6626603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5336-08-3 | C5H8O5 | 148.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5336-08-3
Molecular Formula C5H8O5
Molecular Weight (g/mol) 148.11
MDL Number MFCD00063241
InChI Key CUOKHACJLGPRHD-UHFFFAOYNA-N
Synonym ribonolactone, d-ribono-1,4-lactone, d-ribonolactone, ribono-gamma-lactone, d +-ribonic acid gamma-lactone, d-+-ribono-1,4-lactone, d-ribopentono-1,4-lactone, d-ribono-gamma-lactone, d-+-ribonic acid gamma-lactone, d-ribonic acid-1,4-lactone
PubChem CID 111064
ChEBI CHEBI:74168
IUPAC Name (3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one
SMILES C(C1C(C(C(=O)O1)O)O)O

Specifications

Melting Point 85°C to 87°C
Color White to Yellow
Quantity 1 g
Beilstein 82057
Solubility Information Soluble in water at 100mg/ml
Formula Weight 148.11
Physical Form Powder
Chemical Name or Material D-Ribonic acid-1,4-lactone
EINECSNumber 226-256-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only