missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Raffinose Pentahydrate

Catalog No. p-7082856
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ2106022 100 g Plastic bottle
AAJ2106036 500 g Plastic bottle
AAJ21060A1 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ2106022 Supplier Thermo Fisher Scientific Supplier No. J2106022

CAS: 17629-30-0 | C18H42O21 | 594.513 g/mol

Chemical Identifiers

CAS 17629-30-0
Molecular Formula C18H42O21
Molecular Weight (g/mol) 594.513
MDL Number MFCD00071590
InChI Key BITMAWRCWSHCRW-ORBWWOJFSA-N
Synonym d-+-raffinosepentahydrate
PubChem CID 134129414
IUPAC Name (3 S,4 S,5 R,6 R)-2-[[(2 R,3 S,4 S,5 S)-6-[(2 S,3 S,4 R,5 R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O

Specifications

Boiling Point 78°C to 82°C
Packaging Plastic bottle
Quantity 100 g
Beilstein 4285763
Formula Weight 594.51
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Raffinose Pentahydrate
EINECSNumber 208-146-9
RTECSNumber LZ5851200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only