missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D(+)-Raffinose pentahydrate, 99+%
CAS: 17629-30-0 | C18H32O16·5H2O | 594.52 g/mol
Supplier: Thermo Scientific Chemicals 195675000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| D(+)-Raffinose pentahydrate | |
| 78.0°C to 80.5°C | |
| Authentic | |
| Glass bottle | |
| MFCD00071590 | |
| 17, V,8, 403 | |
| + 105.00 (20.00°C c=10,water,dry basis) | |
| d-+-raffinosepentahydrate | |
| BITMAWRCWSHCRW-ORBWWOJFSA-N | |
| (3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate | |
| 134129414 | |
| 99+% |
| 17629-30-0 | |
| White | |
| 99% min. (ELSD) (HPLC) | |
| C18H32O16·5H2O | |
| 500 g | |
| 14, 8096 | |
| + 105.00 | |
| (10% in water) Clear colorless | |
| C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O | |
| 594.52 | |
| 594.52 | |
| Crystalline Powder |
Chemical Identifiers
| 17629-30-0 | |
| 594.52 | |
| BITMAWRCWSHCRW-ORBWWOJFSA-N | |
| 134129414 | |
| C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O |
| C18H32O16·5H2O | |
| MFCD00071590 | |
| d-+-raffinosepentahydrate | |
| (3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate |
RUO – Research Use Only