missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Raffinose pentahydrate, 99+%

Catalog No. AC195670250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC195670250 25 g Glass bottle
AC195671000 100 g Glass bottle
AC195675000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC195670250 Supplier Thermo Scientific Chemicals Supplier No. 195670250

CAS: 17629-30-0 | C18H32O16·5H2O | 594.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17629-30-0
Molecular Formula C18H32O16·5H2O
Molecular Weight (g/mol) 594.52
MDL Number MFCD00071590
InChI Key BITMAWRCWSHCRW-ORBWWOJFSA-N
Synonym d-+-raffinosepentahydrate
PubChem CID 134129414
IUPAC Name (3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O

Specifications

Melting Point 78.0°C to 80.5°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 17, V,8, 403
Merck Index 14, 8096
Specific Rotation Condition + 105.00 (20.00°C c=10,water,dry basis)
Specific Rotation + 105.00
Solubility Information (10% in water) Clear colorless
Formula Weight 594.52
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Raffinose pentahydrate
Show More Show Less

RUO – Research Use Only