missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Raffinose pentahydrate, 98%, ≥98%, MP Biomedicals™

Catalog No. p-4403664
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10279780 100 g
ICN10279790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10279780 Supplier MP Biomedicals Inc Supplier No. 0210279780
Only null left

Chemical Identifiers

CAS 17629-30-0
Molecular Formula C18H42O21
Molecular Weight (g/mol) 594.513
InChI Key BITMAWRCWSHCRW-ORBWWOJFSA-N
Synonym d-+-raffinosepentahydrate
PubChem CID 134129414
IUPAC Name (3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O

Specifications

Color White
Density 1.465g/mL
Assay Percent Range 0.98
Quantity 100 g
Formula Weight 594.5
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Raffinose pentahydrate, For HPLC analysis