missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(-)-Quinic Acid 98.0+%, TCI America™
Supplier: TCI America Q000925G
Specifications
| D-(-)-Quinic Acid | |
| 166°C | |
| C7H12O6 | |
| 25 g | |
| AAWZDTNXLSGCEK-UHFFFAOYNA-N | |
| 1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid | |
| 6508 | |
| ≥98.0% (T) |
| 77-95-2 | |
| White | |
| MFCD00003864 | |
| quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi | |
| OC1CC(O)(CC(O)C1O)C(O)=O | |
| 192.17 | |
| 192.17 | |
| Crystalline Powder |
Chemical Identifiers
| 77-95-2 | |
| 192.17 | |
| AAWZDTNXLSGCEK-UHFFFAOYNA-N | |
| 6508 | |
| OC1CC(O)(CC(O)C1O)C(O)=O |
| C7H12O6 | |
| MFCD00003864 | |
| quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi | |
| 1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid |
Safety and Handling
RTECSNumber : GU8650000
TSCA : No