missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Quinic Acid 98.0+%, TCI America™

Catalog No. p-7135617
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00095G 5 g
Q000925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00095G Supplier TCI America Supplier No. Q00095G
Only null left

Chemical Identifiers

CAS 77-95-2
Molecular Formula C7H12O6
Molecular Weight (g/mol) 192.17
MDL Number MFCD00003864
InChI Key AAWZDTNXLSGCEK-UHFFFAOYNA-N
Synonym quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi
PubChem CID 6508
IUPAC Name 1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid
SMILES OC1CC(O)(CC(O)C1O)C(O)=O

Specifications

Melting Point 166°C
Color White
Quantity 5 g
Formula Weight 192.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-Quinic Acid
RTECSNumber GU8650000
TSCA No