missing translation for 'onlineSavingsMsg'
Learn More

D-Pyroglutamic Acid 98.0+%, TCI America™

Supplier:  TCI America P13545G

 View more versions of this product

Catalog No. P13545G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

D-Pyroglutamic Acid
162°C
C5H7NO3
5 g
ODHCTXKNWHHXJC-GSVOUGTGSA-N
(2R)-5-oxopyrrolidine-2-carboxylic acid
439685
129.12
Crystalline Powder
4042-36-8
White
MFCD00066212
d-pyroglutamic acid, 5-oxo-d-proline, r-5-oxopyrrolidine-2-carboxylic acid, d-pyr-oh, 2r-5-oxopyrrolidine-2-carboxylic acid, d-proline, 5-oxo, r-+-2-pyrrolidone-5-carboxylic acid, d-5-pyrrolidone-2-carboxylic acid, r-2-pyrrolidone-5-carboxylic acid, d-5-oxo-2-pyrrolidinecarboxylic acid
C1CC(=O)NC1C(=O)O
129.115
CHEBI:16924
≥98.0% (T)

Chemical Identifiers

4042-36-8
129.115
ODHCTXKNWHHXJC-GSVOUGTGSA-N
439685
(2R)-5-oxopyrrolidine-2-carboxylic acid
C5H7NO3
MFCD00066212
d-pyroglutamic acid, 5-oxo-d-proline, r-5-oxopyrrolidine-2-carboxylic acid, d-pyr-oh, 2r-5-oxopyrrolidine-2-carboxylic acid, d-proline, 5-oxo, r-+-2-pyrrolidone-5-carboxylic acid, d-5-pyrrolidone-2-carboxylic acid, r-2-pyrrolidone-5-carboxylic acid, d-5-oxo-2-pyrrolidinecarboxylic acid
CHEBI:16924
C1CC(=O)NC1C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0115

TSCA : No

SDS
Documents

Documents

Promotions

Promotions