missing translation for 'onlineSavingsMsg'
Learn More

D-Pyroglutamic Acid 98.0+%, TCI America™

Catalog No. p-7135095
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13545G 5 g
P135425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13545G Supplier TCI America Supplier No. P13545G
Only null left

Chemical Identifiers

CAS 4042-36-8
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.115
MDL Number MFCD00066212
InChI Key ODHCTXKNWHHXJC-GSVOUGTGSA-N
Synonym d-pyroglutamic acid, 5-oxo-d-proline, r-5-oxopyrrolidine-2-carboxylic acid, d-pyr-oh, 2r-5-oxopyrrolidine-2-carboxylic acid, d-proline, 5-oxo, r-+-2-pyrrolidone-5-carboxylic acid, d-5-pyrrolidone-2-carboxylic acid, r-2-pyrrolidone-5-carboxylic acid, d-5-oxo-2-pyrrolidinecarboxylic acid
PubChem CID 439685
ChEBI CHEBI:16924
IUPAC Name (2R)-5-oxopyrrolidine-2-carboxylic acid
SMILES C1CC(=O)NC1C(=O)O

Specifications

Melting Point 162°C
Color White
Quantity 5 g
Formula Weight 129.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Pyroglutamic Acid
EINECSNumber (5)-0115
TSCA No