missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| D-Prolinol | |
| Yellow | |
| C5H11NO | |
| 25 g | |
| HVVNJUAVDAZWCB-RXMQYKEDSA-N | |
| [(2R)-pyrrolidin-2-yl]methanol | |
| 2724541 | |
| 101.15 | |
| Liquid |
| 68832-13-3 | |
| 76°C | |
| MFCD00064321 | |
| d-prolinol, r-pyrrolidin-2-ylmethanol, 2r-pyrrolidin-2-ylmethanol, r---2-pyrrolidinemethanol, r-prolinol, 2r-pyrrolidin-2-yl methanol, r-1-pyrrolidin-2-yl-methanol, 2-pyrrolidinemethanol, 2r, r---2-hydroxymethyl pyrrolidine | |
| C1CC(NC1)CO | |
| 101.149 | |
| CHEBI:84258 | |
| ≥99.0% (GC,T) |
Chemical Identifiers
| 68832-13-3 | |
| 101.149 | |
| HVVNJUAVDAZWCB-RXMQYKEDSA-N | |
| 2724541 | |
| [(2R)-pyrrolidin-2-yl]methanol |
| C5H11NO | |
| MFCD00064321 | |
| d-prolinol, r-pyrrolidin-2-ylmethanol, 2r-pyrrolidin-2-ylmethanol, r---2-pyrrolidinemethanol, r-prolinol, 2r-pyrrolidin-2-yl methanol, r-1-pyrrolidin-2-yl-methanol, 2-pyrrolidinemethanol, 2r, r---2-hydroxymethyl pyrrolidine | |
| CHEBI:84258 | |
| C1CC(NC1)CO |
Safety and Handling
TSCA : No