missing translation for 'onlineSavingsMsg'
Learn More

D-Prolinol 99.0+%, TCI America™

Catalog No. p-7135046
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
P12741G 1 g
P12745G 5 g
P127425G 25 g
3 options

Catalog No. P12741G

Supplier: TCI America P12741G

Only null left

Chemical Identifiers

CAS 68832-13-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00064321
InChI Key HVVNJUAVDAZWCB-RXMQYKEDSA-N
Synonym d-prolinol, r-pyrrolidin-2-ylmethanol, 2r-pyrrolidin-2-ylmethanol, r---2-pyrrolidinemethanol, r-prolinol, 2r-pyrrolidin-2-yl methanol, r-1-pyrrolidin-2-yl-methanol, 2-pyrrolidinemethanol, 2r, r---2-hydroxymethyl pyrrolidine
PubChem CID 2724541
ChEBI CHEBI:84258
IUPAC Name [(2R)-pyrrolidin-2-yl]methanol
SMILES C1CC(NC1)CO

Specifications

Color Yellow
Boiling Point 76°C
Quantity 1 g
Formula Weight 101.15
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material D-Prolinol