missing translation for 'onlineSavingsMsg'
Learn More

D-Proline methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog No. p-7047018
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1725903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1725903 Supplier Thermo Scientific Chemicals Supplier No. L1725903

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65365-28-8
Molecular Formula C6H12ClNO2
Molecular Weight (g/mol) 165.617
MDL Number MFCD00083685
InChI Key HQEIPVHJHZTMDP-NUBCRITNSA-N
Synonym d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride
PubChem CID 44629857
IUPAC Name methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
SMILES COC(=O)C1CCCN1.Cl

Specifications

Melting Point 69°C to 71°C
Beilstein 4714125
Quantity 1 g
Formula Weight 165.62
Optical Rotation +31° (c=1 in Water)
Percent Purity 99%
Sensitivity Hygroscopic
Chemical Name or Material D-Proline methyl ester hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only